Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Golgin-97 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583719
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583719 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583719 Supplier Invitrogen™ Supplier No. PA583719
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TGAESKITAL EQKEQELQAL IQQLSIDLQK VTAETQEKED VITHLQEKVA SLEKRLEQNL SGEEHLQELL K Highest antigen sequence indentity to the following orthologs: Mouse - 85%, Rat - 80%.

The Golgi apparatus, which participates in glycosylation and transport of proteins and lipids in the secretory pathway, consists of a series of stacked cisternae (flattened membrane sacs). Interactions between the Golgi and microtubules are thought to be important for the reorganization of the Golgi after it fragments during mitosis. The Golgins are a family of proteins, of which the protein encoded by this gene is a member, that are localized to the Golgi. This encoded protein is associated with Sjogren's syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Golgin-97
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GOLGA1
Gene Accession No. Q92805
Gene Alias 0710001G09Rik; 2210418B03Rik; AW107649; gap junction protein, alpha 4, 37kD; Golga1; golgi autoantigen, golgin subfamily a, 1; Golgi97; golgin A1; golgin subfamily A member 1; golgin-97
Gene Symbols GOLGA1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GOLGA1. Recombinant protein control fragment (Product #RP-102964).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2800
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.