Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GOLIM4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569441
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569441 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569441 Supplier Invitrogen™ Supplier No. PA569441
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

The Golgi complex plays a key role in the sorting and modification of proteins exported from the endoplasmic reticulum. The protein encoded by this gene is a type II Golgi-resident protein. It may process proteins synthesized in the rough endoplasmic reticulum and assist in the transport of protein cargo through the Golgi apparatus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GOLIM4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GOLIM4
Gene Accession No. Q5BJK8
Gene Alias 130 kDa golgi-localized phosphoprotein; 3110027H23Rik; cis Golgi-localized calcium-binding protein; decapacitation factor 10; Df10; GIMPc; Golgi integral membrane protein 4; Golgi integral membrane protein, cis; golgi phosphoprotein 4; Golgi phosphoprotein of 130 kDa; golgi-localized phosphoprotein of 130 kDa; GOLIM4; GOLPH4; GPP130; LOW QUALITY PROTEIN: Golgi integral membrane protein 4; P138; type II Golgi membrane protein
Gene Symbols GOLIM4
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human GOLIM4 (aa 480-529).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 310526
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.