Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GOLM1 Antibody (12), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP312533AF750
Description
GOLM1 Monoclonal antibody specifically detects GOLM1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
GOLM1 | |
Monoclonal | |
Alexa Fluor 750 | |
bA379P1.3, FLJ23608, golgi membrane protein 1, Golgi membrane protein GP73, Golgi phosphoprotein 2C9orf155, golgi protein, 73-kD, GOLPH2 FLJ22634, GP73, GP73 chromosome 9 open reading frame 155, PSEC0257 | |
Recombinant Human GOLM1 Protein | |
100 μg | |
Golgi Apparatus Markers, Signal Transduction | |
51280 | |
Store at 4°C in the dark. | |
IgG2a |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
12 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction