Learn More
Invitrogen™ GOLPH2 Polyclonal Antibody

Description
PA5-23076 reacts with GOLM1 in human samples. PA5-23076 has been successfully used in Western Blot and Immunocytochemistry/Immunofluorescence applications. The PA5-23076 immunogen is a genomic peptide made to an internal region of the human GOLM1 protein (within residues 250-400).
Specifications
Specifications
| Antigen | GOLPH2 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.61 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% BSA, 50% glycerol and 0.05% sodium azide |
| Gene | GOLM1 |
| Gene Accession No. | Q8NBJ4, Q91XA2 |
| Gene Alias | 2310001L02Rik; AW125446; bA379P1.3; C9orf155; D030064E01Rik; epididymis luminal protein 46; FLJ22634; FLJ23608; Golgi membrane protein 1; Golgi membrane protein GP73; golgi phosphoprotein 2; golgi protein, 73-kD; GOLM1; Golph2; GOLPH2Ê; GP73; HEL46; PSEC0242; PSEC0257; RP11-379P1.3; UNQ686/PRO1326 |
| Gene Symbols | GOLM1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.