Learn More
Invitrogen™ GOLPH2 Polyclonal Antibody

Description
Immunogen sequence: GKGNVLGNSK SQTPAPSSEV VLDSKRQVEK EETNEIQVVN EEPQRDRLPQ EPGREQVVED RPVGGRGFGG AGELGQTPQV QAALSVSQEN PEMEGPERDQ LVIPDGQEEE QEAAGEGRNQ QKLRGEDDYN MDENEAESE Highest antigen sequence indentity to the following orthologs: Mouse - 46%, Rat - 45%.
Specifications
Specifications
| Antigen | GOLPH2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | GOLM1 |
| Gene Accession No. | Q8NBJ4 |
| Gene Alias | 2310001L02Rik; AW125446; bA379P1.3; C9orf155; D030064E01Rik; epididymis luminal protein 46; FLJ22634; FLJ23608; Golgi membrane protein 1; Golgi membrane protein GP73; golgi phosphoprotein 2; golgi protein, 73-kD; GOLM1; Golph2; GOLPH2Ê; GP73; HEL46; PSEC0242; PSEC0257; RP11-379P1.3; UNQ686/PRO1326 |
| Gene Symbols | GOLM1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.