Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GOLPH2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585235
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585235 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585235 Supplier Invitrogen™ Supplier No. PA585235
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Positive Control: 293T, HeLa, HeLa membrane fraction, DU145. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The Golgi complex plays a key role in the sorting and modification of proteins exported from the endoplasmic reticulum. The protein encoded by this gene is a type II Golgi transmembrane protein. It processes protein synthesized in the rough endoplasmic reticulum and assists in the transport of protein cargo through the Golgi apparatus. The expression of this encoded protein has been observed to be upregulated in response to viral infection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GOLPH2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene GOLM1
Gene Accession No. Q8NBJ4
Gene Alias 2310001L02Rik; AW125446; bA379P1.3; C9orf155; D030064E01Rik; epididymis luminal protein 46; FLJ22634; FLJ23608; Golgi membrane protein 1; Golgi membrane protein GP73; golgi phosphoprotein 2; golgi protein, 73-kD; GOLM1; Golph2; GOLPH2Ê; GP73; HEL46; PSEC0242; PSEC0257; RP11-379P1.3; UNQ686/PRO1326
Gene Symbols GOLM1
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human GOLPH2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51280
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.