Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GOLPH3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589000
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589000 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589000 Supplier Invitrogen™ Supplier No. PA589000
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MTSLTQRSSG LVQRRTEASR NAADKERAAG GGAGSSEDDA QSRRDEQDDD DKGDSKETRL TLMEEVLLLG LKDREGYTSF WNDCISSGLR GCMLIELALR GRLQLEACGM RRKSLLTRKV ICKSDAPTGD VLLDEALKHV KETQPPETVQ NWIELLSGET WNPLKLHYQL RNVRERLAKN LVEKGVLTTE KQNFLLFDMT THPLTNNNIK QRLIKKVQEA VLDKWVNDPH RMDRRLLALI YLAHASDVLE NAFAPLLDEQ YDLATKRVRQ LLDLDPEVEC LKANTNEVLW AVVAAFTK; Positive Samples: 22Rv1, LO2, HeLa, Mouse testis, Rat lung; Cellular Location: Cell membrane, Cytoplasmic side, Endosome, Golgi apparatus, Golgi stack membrane, Mitochondrion intermembrane space, Peripheral membrane protein, trans-Golgi network membrane.

GOLPH3 was initially identified as a peripheral membrane protein localized to the trans-Golgi network, but others reported it to be primarily a mitochondrial protein that regulated the mitochondrial mass through the regulation of the mitochondria-specific phospholipid cardiolipin. GOLPH3 has since been implicated in the target of rapamycin (TOR) signaling pathway. Its overexpression in transfected cells led to and increase in anchorage-independent growth and cell proliferation in vitro. Furthermore, GOLPH3-transfected cells enhanced S6 Kinase activity in response to growth factor stimulation by EGF. Simultaneously, AKT phosphorylation increased in these cells, while these events were abrogated in GOLPH3 siRNA treated cells compared to control cells, indicating the GOLPH3 can enhance signaling through TOR-associated complexes. These results suggest that GOLPH3 is a bona fide oncogene and may be a useful target for therapeutic strategies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GOLPH3
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.87 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Golph3
Gene Accession No. Q9CRA5, Q9ERE4, Q9H4A6
Gene Alias 4733401N08Rik; 5730410D03Rik; AW413496; coat protein GPP34; coat-protein; Gmx33; golgi peripheral membrane protein 1, 34 kDa; Golgi phosphoprotein 3; golgi phosphoprotein 3 (coat-protein); golgi protein; golgi-associated protein; GOLPH3; GOPP1; GPP34; MIDAS; Mitochondrial DNA absence factor; trans-Golgi protein GMx33; Vps74
Gene Symbols Golph3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-298 of human GOLPH3 (NP_0714131).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64083, 66629, 78961
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.