Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GOLPH3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598901
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598901 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598901 Supplier Thermo Scientific Supplier No. PA598901
Add to Cart
Add to Cart

GOLPH3 Polyclonal Antibody

GOLPH3 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IP, ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen GOLPH3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 3.99 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Golph3
Gene Accession No. Q9H4A6
Gene Alias 4733401N08Rik; 5730410D03Rik; AW413496; coat protein GPP34; coat-protein; Gmx33; golgi peripheral membrane protein 1, 34 kDa; Golgi phosphoprotein 3; golgi phosphoprotein 3 (coat-protein); golgi protein; golgi-associated protein; GOLPH3; GOPP1; GPP34; MIDAS; Mitochondrial DNA absence factor; trans-Golgi protein GMx33; Vps74
Gene Symbols Golph3
Host Species Rabbit
Immunogen Recombinant Human Golgi phosphoprotein 3 protein (16-150AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64083
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.