Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GOLPH3L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546472
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546472 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546472 Supplier Invitrogen™ Supplier No. PA546472
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: EISKNSEKKM ESEEDSNWEK SPDNEDSGDS KDIRLTLMEE VLLLGLKDKE Sequence homology: Human: 100%; Mouse: 79%; Rat: 79%.

The Golgi complex plays a key role in the sorting and modification of proteins exported from the endoplasmic reticulum. The protein encoded by this gene is localized at the Golgi stack and may have a regulatory role in Golgi trafficking.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GOLPH3L
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GOLPH3L
Gene Accession No. Q9H4A5
Gene Alias 2010204I15Rik; AU015439; AW549227; C77637; golgi phosphoprotein 3 like; golgi phosphoprotein 3-like; Golph3l; GPP34R; GPP34-related protein
Gene Symbols GOLPH3L
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human GOLPH3L (aa 12-61).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55204
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.