Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GOLT1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA525522
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA525522 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA525522 Supplier Invitrogen™ Supplier No. PA525522
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GOLT1A Polyclonal Antibody for Western Blot, Flow

GOLT1A may be involved in fusion of ER-derived transport vesicles with the Golgi complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GOLT1A
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene GOLT1A
Gene Accession No. Q6ZVE7
Gene Alias 0610012C01Rik; CGI-141; Golgi transport 1 homolog A; golgi transport 1 homolog A (S. cerevisiae); golgi transport 1A; golt1a; GOT1; GOT1B; hGOT1b; HMFN1187; Kiss1; metastatin; putative protein product of HMFN1187; RGD1566310; RP11-203F10.3; Vesicle transport protein GOT1A; YMR292W
Gene Symbols GOLT1A
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 103-130 amino acids from the C-terminal region of human GOT1A.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 127845
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.