Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GOLT1A Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982913
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2913 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2913 Supplier Bioss Supplier No. BS13491R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GOLT1A may be involved in fusion of ER-derived transport vesicles with the Golgi complex.

Specifications

Antigen GOLT1A
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GOLT1A
Gene Accession No. Q6ZVE7
Gene Alias 0610012C01Rik; CGI-141; Golgi transport 1 homolog A; golgi transport 1 homolog A (S. cerevisiae); golgi transport 1A; Golt1a; GOT1; GOT1B; hGOT1b; HMFN1187; Kiss1; metastatin; putative protein product of HMFN1187; RP11-203F10.3; Vesicle transport protein GOT1A; YMR292W
Gene Symbols GOLT1A
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GOLT1A.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 127845
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.