Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GOLT1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565988
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565988 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565988 Supplier Invitrogen™ Supplier No. PA565988
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RRVPVLGSLLN LPGIRSFVDK VGESN Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

GOT1B may be involved in fusion of ER-derived transport vesicles with the Golgi complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GOLT1B
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GOLT1B
Gene Accession No. Q9Y3E0
Gene Alias 2310061A22Rik; AA407874; AU024291; CGI-141; EGK_03451; GCT2; Germ cell tumor 2; Golgi transport 1 homolog B; golgi transport 1 homolog B (S. cerevisiae); golgi transport 1B; Golgi transport 1-like protein B; GOLT1B; GOT1; GOT1A; GOT1B; HDCMA39P; hGOT1a; putative NF-kappa-B-activating protein 470; RGD1310691; UNQ432/PRO793; Vesicle transport protein GOT1B; YMR292W
Gene Symbols GOLT1B
Host Species Rabbit
Immunogen Recombinant Human GOLT1B. Recombinant protein control fragment (Product #RP-103631).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51026
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.