Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GON4L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567350
Description
Immunogen sequence: PVCMDSFQPMD DSLIAFQTRS KMPLKDVPLG QLEAELQAPD ITPDMYDPNT ADDEDWKMWL GGLMNDDVGN EDE Highest antigen sequence identity to the following orthologs - mouse 93%, rat 93%.
Has transcriptional repressor activity, probably as part of a complex with YY1, SIN3A AND HDAC1. Required for B cell lymphopoiesis.
Specifications
GON4L | |
Polyclonal | |
Unconjugated | |
GON4L | |
2610100B20Rik; Dingo; Gon4; GON-4; GON-4 homolog; gon-4 like; GON4L; gon-4-like (C. elegans); GON-4-like protein; KIAA1606; Protein GON4; YARP; YY1AP related protein; YY1AP-related protein1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54856 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q3T8J9 | |
GON4L | |
Recombinant Human GON4L. Recombinant protein control fragment (Product #RP-108004). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction