Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GON4L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568195
Description
GON4L Polyclonal Antibody for Western Blot
Has transcriptional repressor activity, probably as part of a complex with YY1, SIN3A AND HDAC1. Required for B cell lymphopoiesis.
Specifications
GON4L | |
Polyclonal | |
Unconjugated | |
GON4L | |
2610100B20Rik; Dingo; Gon4; GON-4; GON-4 homolog; gon-4 like; GON4L; gon-4-like (C. elegans); GON-4-like protein; KIAA1606; Protein GON4; YARP; YY1AP related protein; YY1AP-related protein1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
54856 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 30% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3T8J9 | |
GON4L | |
Synthesized peptide derived from human GON4L. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction