Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GON4L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103859
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103859 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103859 Supplier Invitrogen™ Supplier No. PA5103859
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GON4L.

Has transcriptional repressor activity, probably as part of a complex with YY1, SIN3A AND HDAC1. Required for B cell lymphopoiesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GON4L
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GON4L
Gene Accession No. Q3T8J9, Q535K8
Gene Alias 2610100B20Rik; Dingo; Gon4; GON-4; GON-4 homolog; gon-4 like; GON4L; gon-4-like (C. elegans); GON-4-like protein; KIAA1606; Protein GON4; YARP; YY1AP related protein; YY1AP-related protein1
Gene Symbols GON4L
Host Species Rabbit
Immunogen A synthesized peptide derived from human GON4L(Accession Q3T8J9), corresponding to amino acid residues K210-E260.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 499653, 54856
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.