Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GON4L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566076
Description
Immunogen sequence: PMEEQDNEESE KRRKKKKGTK RKRDGRGQEG TLAYDLKLDD MLDRTLEDGA KQHNLTAVNV RNILHEVITN EHVVAMMKAA ISETEDMPMF EPKMTRSKLK EVV Highest antigen sequence identity to the following orthologs - mouse 88%, rat 90%.
Has transcriptional repressor activity, probably as part of a complex with YY1, SIN3A AND HDAC1. Required for B cell lymphopoiesis.
Specifications
GON4L | |
Polyclonal | |
Unconjugated | |
GON4L | |
2610100B20Rik; Dingo; Gon4; GON-4; GON-4 homolog; gon-4 like; GON4L; gon-4-like (C. elegans); GON-4-like protein; KIAA1606; Protein GON4; YARP; YY1AP related protein; YY1AP-related protein1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54856 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q3T8J9 | |
GON4L | |
Recombinant Human GON4L. Recombinant protein control fragment (Product #RP-106768). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction