Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Goosecoid Antibody (OTI1D7), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272409MFV500
Description
Goosecoid Monoclonal antibody specifically detects Goosecoid in Human, Mouse, Rat samples. It is validated for Flow CytometrySpecifications
Goosecoid | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Flow Cytometry | |
OTI1D7 | |
Flow Cytometry | |
goosecoid homeobox, homeobox protein goosecoid | |
Human recombinant protein fragment corresponding to amino acids 107-257 of human GSC (NP_776248) produced in E.coli. | |
0.1 mL | |
Stem Cells | |
145258 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction