Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Goosecoid Antibody (SR1466), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $413.50
Specifications
Antigen | Goosecoid |
---|---|
Clone | SR1466 |
Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1:50 |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Description
Goosecoid Monoclonal antibody specifically detects Goosecoid in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
Goosecoid | |
Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
goosecoid homeobox, homeobox protein goosecoid | |
A synthesized peptide derived from human Goosecoid (Uniprot #: P56915) | |
Primary | |
Store at -20°C. Avoid freeze/thaw cycles. |
SR1466 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Stem Cells | |
PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
145258 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title