Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goosecoid Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501993161
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3161 100 μL
50-199-3129 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3161 Supplier Active Motif Supplier No. 61121
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a member of the bicoid subfamily of the paired (PRD) homeobox family of proteins. The encoded protein acts as a transcription factor and may be autoregulatory. A similar protein in mice plays a role in craniofacial and rib cage development during embryogenesis.

Specifications

Antigen Goosecoid
Applications Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
Gene GSC
Gene Accession No. P56915
Gene Alias BOS_20627; GCS; goosecoid; goosecoid homeobox; goosecoid protein; Gsc; Homeobox protein goosecoid; SAMS
Gene Symbols GSC
Host Species Rabbit
Immunogen This Goosecoid antibody was raised against a peptide in the N-terminal region of human homeobox protein Goosecoid.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 145258
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.