Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goosecoid Rabbit anti-Human, Mouse, Clone: 6M7M6, Novus Biologicals™
SDP

Catalog No. p-7247434 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166970 20 μg
NB166969 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB166970 Supplier Novus Biologicals Supplier No. NBP31593720UL
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Goosecoid Monoclonal antibody specifically detects Goosecoid in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen Goosecoid
Applications Western Blot
Classification Monoclonal
Clone 6M7M6
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias goosecoid homeobox, homeobox protein goosecoid
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Goosecoid (P56915).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 145258
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.