Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GORAB Monoclonal Antibody (OTI4A4), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA505038S


Description
GORAB Monoclonal antibody specifically detects GORAB in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
GORAB | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P35250 | |
Mouse | |
Affinity chromatography | |
RUO | |
5982 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Western Blot | |
OTI4A4 | |
Unconjugated | |
RFC2 | |
AI467484; dynactin 1, isoform 1; dynactin 1 (p150, Glued homolog); p150, Glued homolog; 150 kDa dynein-associated polypeptide; p150-glued; GO; golgin, RAB6 interacting; golgin, RAB6-interacting; Gorab; hNTKL-BP1; mNTKL-BP1; N-terminal kinase-like-binding protein 1; NTKL-binding protein 1; NTKLBP1; NTKL-BP1; RAB6-interacting golgin; RAB6-interacting golgin-like protein; SCY1-like 1 binding protein 1; SCY1-like 1-binding protein 1; SCYL1-binding protein 1; Scyl1bp1; SCYL1-BP1 | |
Full length human recombinant protein of human RFC2 produced in E.coli. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction