Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GORAB Monoclonal Antibody (OTI4A4), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA505038

Description
GORAB encodes a member of the golgin family, a group of coiled-coil proteins localized to the Golgi. The encoded protein may function in the secretory pathway. The encoded protein, which also localizes to the cytoplasm, was identified by interactions with the N-terminal kinase-like protein, and thus it may function in mitosis.Specifications
GORAB | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q5T7V8 | |
GORAB | |
Human recombit protein fragment corresponding to amino acids 35-394 of human GORAB produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Western Blot | |
OTI4A4 | |
Unconjugated | |
GORAB | |
GO, NTKLBP1, SCYL1BP1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
92344 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction