Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GORAB Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA551108

Catalog No. PA551108


Only null left
Add to Cart

Description

Description

GORAB Polyclonal Antibody detects endogenous levels of GORAB protein.

TCEB1 is known as protein elongin C, which is a subunit of the transcription factor B (SIII) complex. The SIII complex is composed of elongins A/A2, B and C. It activates elongation by RNA polymerase II by suppressing transient pausing of the polymerase at many sites within transcription units. Elongin A functions as the transcriptionally active component of the SIII complex, whereas elongins B and C are regulatory subunits. Elongin A2 is specifically expressed in the testis, and capable of forming a stable complex with elongins B and C. The von Hippel-Lindau tumor suppressor protein binds to elongins B and C, and thereby inhibits transcription elongation. Western blots using two different antibodies (P100962-T200 and P100963_P050 / P100963_P100) against two unique regions of this protein target confirm the same apparent molecular weight in our tests.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GORAB
Polyclonal
Unconjugated
Gorab
AI467484; dynactin 1, isoform 1; dynactin 1 (p150, Glued homolog); p150, Glued homolog; 150 kDa dynein-associated polypeptide; p150-glued; GO; golgin, RAB6 interacting; golgin, RAB6-interacting; Gorab; hNTKL-BP1; mNTKL-BP1; N-terminal kinase-like-binding protein 1; NTKL-binding protein 1; NTKLBP1; NTKL-BP1; RAB6-interacting golgin; RAB6-interacting golgin-like protein; SCY1-like 1 binding protein 1; SCY1-like 1-binding protein 1; SCYL1-binding protein 1; Scyl1bp1; SCYL1-BP1
Rabbit
Antigen affinity chromatography
RUO
304923, 92344, 98376
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
B1H222, Q5T7V8, Q8BRM2
Gorab
Synthesized peptide derived from the N-terminal region of human SCYL1BP1.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.