Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GORAB Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
| Antigen | GORAB |
|---|---|
| Applications | Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Liquid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-1027
|
Bethyl Laboratories, Inc
A305691AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
GORAB encodes a member of the golgin family, a group of coiled-coil proteins localized to the Golgi. The encoded protein may function in the secretory pathway. The encoded protein, which also localizes to the cytoplasm, was identified by interactions with the N-terminal kinase-like protein, and thus it may function in mitosis.Specifications
| GORAB | |
| Polyclonal | |
| Liquid | |
| RUO | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| GO, NTKLBP1, SCYL1BP1, N-terminal kinase-like-binding protein 1, NTKL-binding protein 1, RAB6-interacting golgin, SCY1-like 1 binding protein 1, SCY1-like 1-binding protein 1, SCYL1-BP1, SCYL1-binding protein 1, hNTKL-BP1 | |
| GORAB | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
| Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q5T7V8 | |
| 92344 | |
| Between 75 and 125 | |
| Primary | |
| 4° C | |
| GORAB |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title