Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GORASP2 Monoclonal Antibody (1C9A3), CoraLite488, Proteintech
SDP

Catalog No. 502334589 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-233-4589 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-233-4589 Supplier Proteintech Group Inc Supplier No. CL48866627100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GORASP2 Monoclonal antibody specifically detects GORASP2 in Human, Pig, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry

Specifications

Antigen GORASP2
Applications Immunocytochemistry
Classification Monoclonal
Clone 1C9A3
Concentration 1 mg/mL
Conjugate CoraLite 488
Formulation PBS with 0.5% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene Accession No. Q9H8Y8
Gene Alias GOLPH6; GORASP2; GRASP55; GRS2; p59
Host Species Mouse
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26003
Target Species Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.