Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GORASP2 Mouse anti-Human, Porcine, Rat, Clone: 1C9A3, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | GORASP2 |
---|---|
Clone | 1C9A3 |
Concentration | 1 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-7045
![]() ![]() |
Proteintech Group Inc
666271IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
GRASP55 is a PDZ containing protein involved in the stacking of golgi cisternea in a cell-free system.Specifications
GORASP2 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Golgi phosphoprotein 6, GOLPH6, GORASP2, GRASP55, GRS2, p59 | |
Gorasp2 | |
IgG1 | |
Protein G | |
Antibody |
1C9A3 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Pig, Rat | |
Q9H8Y8, Q9R064 | |
100154960, 113961, 26003 | |
GORASP2 Fusion Protein Ag17607 | |
Primary | |
-20°C | |
GORASP2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title