Learn More
Promega GoScript Reverse Transcription Mix, Random Primers, 50 reactions
Catalog No. PRA2800 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
50 reactions
Catalog No. Quantity
PRA2800 50 reactions
1 options
Catalog No. PRA2800 Supplier Promega Supplier No. A2800
GoScript Reverse Transcriptase utilizes M-MLV and state-of-the-art buffer technology designed for qPCR to deliver robust, reliable cDNA synthesis of a full range of rare and abundant transcripts, even in the presence of inhibitors.
  • Reliable cDNA Synthesis for Sensitive Detection of Expression
  • Available as a standalone enzyme, a complete reverse transcription kit or as a master mix with Oligo(dT) or Random Primers. RNasin(R) RNase Inhibitor protection included with Kit and Mixes
  • Sensitive transcription of both high−copy and low−copy messages
  • Transcribes short and long transcripts; processes through secondary structure
Specifications
Content And Storage -30C TO -10C
Quantity 50 reactions