Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GOSR1 Antibody (HFD9), Novus Biologicals™
SDP

Catalog No. NBP19743702 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
0.2 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP19743702 0.2 mg
NB018761 0.025 mg
NBP19743705 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP19743702 Supplier Novus Biologicals Supplier No. NBP1974370.2MG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GOSR1 Monoclonal specifically detects GOSR1 in Mouse, Rat, Porcine, Bovine, Canine, Chicken, Rabbit, Sheep samples. It is validated for Western Blot, Electron Microscopy.

Specifications

Antigen GOSR1
Applications Western Blot, Immunoassay
Classification Monoclonal
Clone HFD9
Conjugate Unconjugated
Dilution Western Blot 1:1000/ECL, Electron Microscopy 1:10-1:500
Formulation PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide
Gene Alias golgi integral membrane protein 2, golgi SNAP receptor complex member 1,28 kDa cis-Golgi SNARE p28, Golgi SNARE 28 kDa, GOLIM2, GOS28cis-golgi SNARE, GOS-28GOS28/P28, GS2828 kDa Golgi SNARE protein, P28
Gene Symbols GOSR1
Host Species Mouse
Immunogen Rat liver Golgi-enriched membranes.
Purification Method Protein G purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 9527
Target Species Mouse, Rat, Pig, Bovine, Canine, Chicken, Rabbit, Sheep
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.