Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GOSR1 Monoclonal Antibody (HFD9-5)
GREENER_CHOICE

Catalog No. PIMA191008
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA191008 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA191008 Supplier Invitrogen™ Supplier No. MA191008
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Recommended positive controls: Rat and mouse brain tissue extract.

This gene encodes a trafficking membrane protein which transports proteins among the endoplasmic reticulum and the Golgi and between Golgi compartments. This protein is considered an essential component of the Golgi SNAP receptor (SNARE) complex. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GOSR1
Applications Immunomicroscopy, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone HFD9-5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.1mM PMSF and 0.09% sodium azide; pH 7.2
Gene GOSR1
Gene Accession No. O08522, O88630, Q2TBU3, Q62931
Gene Alias 28 kDa cis-Golgi SNARE p28; 28 kDa Golgi SNARE protein; AI414660; AI426320; BB145494; cis-Golgi p28 (p28); cis-golgi SNARE; golgi integral membrane protein 2; golgi SNAP receptor complex member 1; Golgi SNARE 28 kDa; GOLIM2; Gos28; GOS-28; GOS28/P28; Gosr1; GOSRI; Gs28; hypothetical protein LOC511030; hypothetical protein LOC550344; I79_013357; p28; RP23-343C18.3; v-SNARE; zgc:112064
Gene Symbols GOSR1
Host Species Mouse
Immunogen Rat liver Golgi-enriched membranes.
Purification Method Protein G
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100339588, 100517676, 100689312, 101112332, 491185, 511030, 53334, 94189
Target Species Mouse, Rat, Canine, Rabbit, Bovine, Sheep, Hamster, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.