Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GOSR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544994
Description
Peptide sequence: IHSKMNTLAN RFPAVNSLIQ RINLRKRRDS LILGGVIGIC TILLLLYAFH Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
This gene encodes a trafficking membrane protein which transports proteins among the endoplasmic reticulum and the Golgi and between Golgi compartments. This protein is considered an essential component of the Golgi SNAP receptor (SNARE) complex. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
Specifications
| GOSR1 | |
| Polyclonal | |
| Unconjugated | |
| GOSR1 | |
| 28 kDa cis-Golgi SNARE p28; 28 kDa Golgi SNARE protein; AI414660; AI426320; BB145494; cis-Golgi p28 (p28); cis-golgi SNARE; golgi integral membrane protein 2; golgi SNAP receptor complex member 1; Golgi SNARE 28 kDa; GOLIM2; Gos28; GOS-28; GOS28/P28; Gosr1; GOSRI; Gs28; hypothetical protein LOC511030; hypothetical protein LOC550344; I79_013357; p28; RP23-343C18.3; v-SNARE; zgc:112064 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 9527 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O95249 | |
| GOSR1 | |
| Synthetic peptide directed towards the C-terminal of human GOSR1 (aa 201-250). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction