Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GOSR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599634
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599634 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599634 Supplier Invitrogen™ Supplier No. PA599634
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GS28.

This gene encodes a trafficking membrane protein which transports proteins among the endoplasmic reticulum and the Golgi and between Golgi compartments. This protein is considered an essential component of the Golgi SNAP receptor (SNARE) complex. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GOSR1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GOSR1
Gene Accession No. O88630, O95249
Gene Alias 28 kDa cis-Golgi SNARE p28; 28 kDa Golgi SNARE protein; AI414660; AI426320; BB145494; cis-Golgi p28 (p28); cis-golgi SNARE; golgi integral membrane protein 2; golgi SNAP receptor complex member 1; Golgi SNARE 28 kDa; GOLIM2; Gos28; GOS-28; GOS28/P28; Gosr1; GOSRI; Gs28; hypothetical protein LOC511030; hypothetical protein LOC550344; I79_013357; p28; RP23-343C18.3; v-SNARE; zgc:112064
Gene Symbols GOSR1
Host Species Rabbit
Immunogen A synthesized peptide derived from human GOSR1(Accession O95249), corresponding to amino acid residues K126-L176.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53334, 9527
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.