Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GOSR2/Membrin Mouse anti-Human, Clone: 7E7C7, Proteintech
SDP

Catalog No. 501736570 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6570 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6570 Supplier Proteintech Group Inc Supplier No. 661341IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This gene encodes a trafficking membrane protein which transports proteins among the medial- and trans-Golgi compartments. Due to its chromosomal location and trafficking function, this gene may be involved in familial essential hypertension. Three transcript variants encoding three different isoforms have been found for this gene.

Specifications

Antigen GOSR2/Membrin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 7E7C7
Concentration 1.11 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GOSR2
Gene Accession No. O14653
Gene Alias 27 kDa Golgi SNARE protein, Bos1, GOSR2, GS27, Membrin
Gene Symbols GOSR2
Host Species Mouse
Immunogen GOSR2/Membrin Fusion Protein Ag21393
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9570
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.