Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GOSR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562971
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562971 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562971 Supplier Invitrogen™ Supplier No. PA562971
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLSRTFTTND SDTTIPMDES LQFNSSLQKV HNGMDDLILD GHNILDGLRT QRLTLKGTQK KILDIANMLG LSNTVMRLIE KRAFQ Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 91%.

This gene encodes a trafficking membrane protein which transports proteins among the medial- and trans-Golgi compartments. Due to its chromosomal location and trafficking function, this gene may be involved in familial essential hypertension. Three transcript variants encoding three different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GOSR2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Gosr2
Gene Accession No. O14653
Gene Alias 2310032N09Rik; 27 kDa Golgi SNARE protein; Bos1; C76855; EPM6; Golgi SNAP receptor complex member 2; golgi SNARE; Gosr2; Gs27; I79_025823; LOC100766577; Membrin; RP23-272P17.5; SNARE
Gene Symbols Gosr2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GOSR2. Recombinant protein control fragment (Product #RP-101174).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9570
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.