Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GOSR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597799
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597799 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597799 Supplier Invitrogen™ Supplier No. PA597799
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GOSR2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene encodes a trafficking membrane protein which transports proteins among the medial- and trans-Golgi compartments. Due to its chromosomal location and trafficking function, this gene may be involved in familial essential hypertension. Three transcript variants encoding three different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GOSR2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Gosr2
Gene Accession No. O14653
Gene Alias 2310032N09Rik; 27 kDa Golgi SNARE protein; Bos1; C76855; EPM6; Golgi SNAP receptor complex member 2; golgi SNARE; Gosr2; Gs27; I79_025823; LOC100766577; Membrin; RP23-272P17.5; SNARE
Gene Symbols Gosr2
Host Species Rabbit
Immunogen Recombinant Human Golgi SNAP receptor complex member 2 protein (1-190AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9570
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.