Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Corning™ Autoclave Bags Printed with Biohazard Logo
Shop All Corning Products
Click to view available options
Capacity (Metric):
3 L
9 L
14 L
33 L
Width (Metric):
255 mm
300 mm
400 mm
600 mm
Description
- Polypropylene (PP) resistant bags
- Thermal resistance 134°C
- Transparent

Specifications
Specifications
Capacity (Metric) | 3 L |
Material | Polypropylene |
Thickness (Metric) | 40 μm |
No. per Case | 500/Cs. |
No. per Pack | 1/Pk. |
Width (Metric) | 255 mm |
Height (Metric) | 400 mm |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction