Learn More
Invitrogen™ GOT1 Monoclonal Antibody (GT1255)

Description
Positive Control: 293T, A431, HeLa, HepG2, mouse heart, mouse liver Predicted Reactivity: Rat (87%), Zebrafish (81%), Dog (100%), Pig (97%), Chicken (81%), Chimpanzee (100%), Bovine (97%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | GOT1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | GT1255 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and no preservative; pH 7 |
| Gene | GOT1 |
| Gene Accession No. | P05201, P17174 |
| Gene Alias | AI789014; aspartate aminotransferase 1; aspartate aminotransferase, cytoplasmic; aspartate transaminase 1; Aspat; AST1; ASTQTL1; cAspAT; cCAT; Cysteine aminotransferase, cytoplasmic; cysteine transaminase, cytoplasmic; cytosolic aspartate aminotransferase; Gaspat; GIG18; Glutamate oxaloacetate transaminase 1; glutamate oxaloacetate transaminase 1, soluble; glutamic-oxaloacetic transaminase 1; glutamic-oxaloacetic transaminase 1, soluble; glutamic-oxaloacetic transaminase 1, soluble (aspartate aminotransferase 1); GOT1; Got-1; growth-inhibiting protein 18; testis secretory sperm-binding protein Li 196a; transaminase A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.