Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GOT2 Antibody (8B11), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259428MFV450
Description
GOT2 Monoclonal antibody specifically detects GOT2 in Human, Rat samples. It is validated for Flow CytometrySpecifications
GOT2 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Flow Cytometry | |
8B11 | |
Flow Cytometry | |
aspartate aminotransferase, mitochondrial, EC 2.6.1, EC 2.6.1.1, FABP-1, FABPpm, Fatty acid-binding protein, FLJ40994, Glutamate oxaloacetate transaminase 2, glutamic-oxaloacetic transaminase 2, mitochondrial (aspartate aminotransferase2), KAT4, KATIV, kynurenine aminotransferase IV, mAspAT, mitAAT, Plasma membrane-associated fatty acid-binding protein, Transaminase A | |
Recombinant human GOT2 (30-430aa) purified from E.coli. | |
0.1 mL | |
Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction | |
2806 | |
Store at 4C in the dark. | |
IgG3 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction