Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GOT2 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB10093404
Description
GOT2 Polyclonal specifically detects GOT2 in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
GOT2 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
aspartate aminotransferase, mitochondrial, EC 2.6.1, EC 2.6.1.1, FABP-1, FABPpm, Fatty acid-binding protein, FLJ40994, Glutamate oxaloacetate transaminase 2, glutamic-oxaloacetic transaminase 2, mitochondrial (aspartate aminotransferase2), KAT4, KATIV, kynurenine aminotransferase IV, mAspAT, mitAAT, Plasma membrane-associated fatty acid-binding protein, Transaminase A | |
Goat | |
Affinity Purified | |
RUO | |
2806 | |
Human | |
IgG |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 0.03 - 0.1 ug/ml, Peptide ELISA Detection limit 1:32000 | |
NP_002071.2 | |
GOT2 | |
Peptide with sequence CKDADEAKRVES corresponding to internal region according to NP_002071.2. | |
0.1 mg | |
Primary | |
Cross reacts with Human. Expected from sequence similarity: Mouse and Rat. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction