Learn More
GOT2 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304356A

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to react with Rat, X. tropicalis, Chicken, Bovine, Horse, Rabbit, Pig and Orangutan
Glutamic-oxaloacetic transaminase is a pyridoxal phosphate-dependent enzyme which exists in cytoplasmic and inner-membrane mitochondrial forms, GOT1 and GOT2, respectively. GOT plays a role in amino acid metabolism and the urea and tricarboxylic acid cycles. The two enzymes are homodimeric and show close homology.Specifications
GOT2 | |
Polyclonal | |
Unconjugated | |
GOT2 | |
GOT2; glutamic-oxaloacetic transaminase 2, mitochondrial (aspartate aminotransferase 2); FLJ40994; aspartate aminotransferase 2; KAT4; KATIV; mitAAT; kynurenine aminotransferase IV | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14719, 2806 | |
4° C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
P00505, P05202 | |
GOT2 | |
Between 380 and 430 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.