Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GOT2 Recombinant Rabbit Monoclonal Antibody (JE54-46)
GREENER_CHOICE

Catalog No. PIMA536188
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA536188 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA536188 Supplier Invitrogen™ Supplier No. MA536188
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

GOT2 Recombinant Monoclonal Antibody for Flow, IHC (P), Western Blot

GOT2 is a glutamic-oxaloacetic transaminase is a pyridoxal phosphate-dependent enzyme which exists in cytoplasmic and inner-membrane mitochondrial forms, GOT1 and GOT2, respectively. GOT plays a role in amino acid metabolism and the urea and tricarboxylic acid cycles. The two enzymes are homodimeric and show close homology.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GOT2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JE54-46
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene GOT2
Gene Accession No. P00505, P00507, P05202
Gene Alias AL022787; aspartate aminotransferase 2; aspartate aminotransferase precursor (EC 2.6.1.1); aspartate aminotransferase, mitochondrial; aspartate transaminase 2; ASPATA; FABP-1; FABPpm; FABP-pm; fatty acid-binding protein; FLJ40994; Glutamate oxaloacetate transaminase 2; glutamate oxaloacetate transaminase 2, mitochondrial; Glutamate oxaloacetate transaminase 2, mitochondrial (aspartate aminotransferase 2); glutamatic-oxaloacetic transaminase 2, mitochondrial; glutamic-oxaloacetic transaminase 2; glutamic-oxaloacetic transaminase 2, mitochondrial; glutamic-oxaloacetic transaminase 2, mitochondrial (aspartate aminotransferase 2); GOT2; Got-2; KAT4; KATIV; KYAT4; kynurenine aminotransferase 4; Kynurenine aminotransferase IV; kynurenine--oxoglutarate transaminase 4; Kynurenine--oxoglutarate transaminase IV; Maat; mAspAT; mitAAT; mitochondrial aspartate aminotransferase; plasma membrane fatty acid binding protein; plasma membrane-associated fatty acid-binding protein; Transaminase A
Gene Symbols GOT2
Host Species Rabbit
Immunogen Recombinant protein within Human GOT2 aa 1-139.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14719, 25721, 2806
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.