Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega GoTaq™ Flexi DNA Polymerase

Catalog No. p-2902615 Shop All Promega Corporation Products
Change view
Click to view available options
Description:
DNA Polymerase
Quantity:
10000U
100U
2500U
5000U
500U
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Quantity
PR-M8291 DNA Polymerase 100U
PR-M8297 DNA Polymerase 5000U
PR-M8296 DNA Polymerase 2500U
PRM8298 DNA Polymerase 10000U
PR-M8295 DNA Polymerase 500U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. PR-M8291 Supplier Promega Supplier No. M8291
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

GoTaq Flexi DNA Polymerase, 5X Colorless GoTaq Flexi Buffer (Used when direct fluorescence or absorbance readings are required without prior purification of the DNA from the amplification reaction), 5X Green GoTaq Flexi Buffer (Contains two dyes that separate during electrophoresis to facilitate direct-to-gel analysis of PCR products), 25mM MgCl2

Taq DNA Polymerase. Provided with Green and Colorless Reaction Buffers without MgCl2, and 25mM MgCl2. Green Buffer allows direct gel analysis after amplification. Enhanced performance over standard Taq DNA Polymerase.

  • Optimize Enzyme and Magnesium Concentrations in Your PCR
  • Mg-free reaction buffers for ease of optimization
  • Direct-to-gel convenience
  • Supplied at a concentration of 5u/μL

  • The purchase of this product conveys to the buyer the limited, nonexclusive, nontransferable right (without the right to resell, repackage, or further sublicense) under U.S. Published Patent Appln. 20020150900 and U.S. Pat. Nos. 5,432,272, 6,617,106 and 6,140,496 to use the product. No other license is granted to the buyer whether expressly, by implication, by estoppel or otherwise. In particular, the purchase of this product does not include or carry any right or license to sell this product. For information on purchasing a license for other uses, please contact Promega Corporation, Licensing, 2800 Woods Hollow Road, Madison, WI 53711, or EraGen Biosciences, Corporate Licensing, 918 Deming Way, Suite 201, Madison, WI 53717. Phone (608) 662-9000; Fax (608) 662-9003.
  • This product is designed and sold for use in the multiplex PCR process covered by U.S. Pat. No. 5,582,989 and Canadian Pat. No. 1,339,731. A limited license has been granted under the patent to use only this amount of the product to practice the multiplex PCR process and is conveyed to the purchaser by the purchase of this product.
  • Patents for the foundational PCR process, European Pat. Nos. 201,184 and 200,362, expired on March 28, 2006. In the U.S., the patents covering the foundational PCR process expired on March 29, 2005.

Specifications

Concentration 5 U/μL
Content And Storage −30 to −10°C
Description DNA Polymerase
Form Liquid
pH 8
Quantity 100U
For Use With (Application) Standard PCR
Includes GoTaq Flexi DNA Polymerase, 5X Colorless GoTaq Flexi buffer (Used when direct fluorescence or absorbance readings are required without prior purification of the DNA from the amplification reaction), 5X Green GoTaq Flexi buffer, 25mM MgCl2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.