Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega GoTaq™ G2 Hot Start Polymerase

Catalog No. PRM7408 Shop All Promega Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
10,000U
100U
2500U
500U
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRM7408 10,000U
PRM7401 100U
PRM7405 500U
PRM7406 2500U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. PRM7408 Supplier Promega Supplier No. M7408
Add to Cart
Add to Cart
Request a Quote

GoTaq™ G2 Hot Start Polymerase is supplied with 5X Green GoTaq™ Flexi Buffer, 5X Colorless GoTaq™ Flexi Buffer and 25mM MgCl2.

  • High Performance GoTaq(R) G2 Enzyme with Convenient Room-Temperature Setup
  • Eliminate nonspecific amplification with a hot start enzyme
  • Detect lower-copy-number targets
  • Optimize reaction conditions with a Flexi Reaction Buffer and separate MgCl2
  • Supplied at a concentration of 5u/μL

Specifications

Concentration 5 U/μL
Content And Storage −30° to −10°C
Polymerase GoTaq G2
Quantity 10,000U
Sample Type Hot Start Polymerase
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.