Learn More
Promega GoTaq MDx DNA Polymerase, 500u
Catalog No. PRD4005 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
500u
Catalog No. Quantity
PRD4005 500u
1 options
This item is not returnable. View return policy
Catalog No. PRD4005 Supplier Promega Supplier No. D4005
This item is not returnable. View return policy
GoTaq MDx DNA Polymerase is a full-length form of Taq DNA polymerase that exhibits 5'→3' exonuclease activity.
  • Full−Length Form of Taq DNA Polymerase Manufactured Under cGMP
  • Glycerol−free formulation available
  • Custom formats and formulations
  • Supplied at a concentration ≥5u/microl
Specifications
Content And Storage -30C TO -10C
Quantity 500u