Learn More
Promega 5X Green GoTaq™ Reaction Buffer
Catalog No. PRM7911 Shop All Promega Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
20mL
Catalog No. Quantity
PRM7911 20mL
1 options
Catalog No. PRM7911 Supplier Promega Supplier No. M7911
The 5X Green GoTaq Reaction Buffer contains a compound that increases sample density as well as two dyes that separate during electrophoresis to show migration progress. Samples can be loaded directly onto gels without the need for loading dye.
  • Buffers for Reliable, Robust PCR
  • Available with green dye for direct-to-gel convenience, or in colorless format for applications where absorbance or fluorescence measurements are necessary prior to PCR cleanup
  • Contain 7.5mM MgCl2 for a final concentration of 1.5mM in a 1X reaction
Specifications
Quantity 20mL