Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GP-IX Antibody (GR-P) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP309133
Description
GP-IX Monoclonal specifically detects GP-IX in Human, Canine samples. It is validated for Flow Cytometry.Specifications
| GP-IX | |
| Monoclonal | |
| 1 mg/ml | |
| Flow Cytometry 1-4 ug/ml | |
| CD42a, CD42a antigen, Glycoprotein 9, glycoprotein IX (platelet), GP-IX, GPIXplatelet glycoprotein IX | |
| Human acute lymphoblastic leukemia cells | |
| 0.1 mg | |
| Primary | |
| Human, Canine | |
| Purified |
| Flow Cytometry | |
| GR-P | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 2815 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction