Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GP repeat Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5141023
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5141023 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5141023 Supplier Invitrogen™ Supplier No. PA5141023
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GP repeat Polyclonal Antibody for Western Blot

The C9orf72 GGGGCC repeat sequence codes five repeat peptide GA repeat; GAGAGAGAGA, GP repeat; GPGPGPGPG, GR repeat; GRGRGRGRG, AP repeat; APAPAPAPA and PR repeat; PRPRPRPRP. It was described previously that aggregated forms of poly-GA and poly-GP proteins do not enter the separation gel.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GP repeat
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene Alias C9RAN; GGGCC; GP repeat; GPGPGPGP; poly GP; poly PG
Host Species Rabbit
Immunogen Synthetic peptide immune sequence is GPGPGPGPGC.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.