Learn More
GP repeat Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 244941AP150UL

Description
This antibody is used to detect the GP repeated sequence. The recombit protein 6xHis tag-SMARCA4 or GST tag-SMARCA4 which contains the sequence 'GPGPGPGPG' were used to detect this antibody.
The C9orf72 "GGGGCC" repeat sequence codes five repeat peptide "GA repeat; GAGAGAGAGA", "GP repeat; GPGPGPGPG", "GR repeat; GRGRGRGRG", "AP repeat; APAPAPAPA" and "PR repeat; PRPRPRPRP". It was described previously that aggregated forms of poly-GA and poly-GP proteins do not enter the separation gel.Specifications
GP repeat | |
Polyclonal | |
Unconjugated | |
C9RAN, GGGCC, GP repeat, GPGPGPGP, poly GP, poly PG | |
Peptide | |
150 μL | |
Primary | |
-20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.