Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
gp130/CD130 Antibody (108), Alexa Fluor™ 750

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290852AF750
Description
gp130/CD130 Monoclonal specifically detects gp130/CD130 in Rat samples. It is validated for Flow Cytometry, ELISA.Specifications
gp130/CD130 | |
Monoclonal | |
Alexa Fluor 750 | |
CD130, CD130 antigen, CDw130, DKFZp564F053, gp130, gp130 of the rheumatoid arthritis antigenic peptide-bearing soluble form, IL-6 receptor subunit beta, IL-6R subunit beta, IL-6RB, IL-6R-beta, interleukin 6 signal transducer (gp130, oncostatin M receptor), interleukin receptor beta chain, interleukin-6 receptor subunit beta, Interleukin-6 signal transducer, Membrane glycoprotein 130, membrane glycoprotein gp130, Oncostatin-M receptor subunit alpha | |
This antibody was obtained from a rabbit immunized with purified, recombinant Rat gp130/CD130 (NP_001008725.2; Met4-Glu618). | |
0.1 mL | |
Cancer, Immunology | |
3572 | |
Store at 4°C in the dark. | |
IgG |
Flow Cytometry, ELISA | |
108 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction