Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
gp130/CD130 Antibody (MM0318-9X23) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50
Specifications
Antigen | gp130/CD130 |
---|---|
Clone | MM0318-9X23 |
Concentration | LYOPH |
Applications | Western Blot, Flow Cytometry, CyTOF, Blocking Assay |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB019185
![]() |
Novus Biologicals
NBP2123440.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NBP212344
![]() |
Novus Biologicals
NBP212344 |
0.1 mg | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
gp130/CD130 Monoclonal specifically detects gp130/CD130 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready, Block/Neutralize.Specifications
gp130/CD130 | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD130, CD130 antigen, CDw130, DKFZp564F053, gp130, gp130 of the rheumatoid arthritis antigenic peptide-bearing soluble form, IL-6 receptor subunit beta, IL-6R subunit beta, IL-6RB, IL-6R-beta, interleukin 6 signal transducer (gp130, oncostatin M receptor), interleukin receptor beta chain, interleukin-6 receptor subunit beta, Interleukin-6 signal transducer, Membrane glycoprotein 130, membrane glycoprotein gp130, Oncostatin-M receptor subunit alpha | |
IL6ST | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
MM0318-9X23 | |
Western Blot, Flow Cytometry, CyTOF, Blocking Assay | |
Unconjugated | |
Mouse | |
Cancer, Immunology | |
P40189 | |
3572 | |
IgG1 | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title