Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
gp130/CD130 Antibody (MM0318-9X23), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212344PCP
Description
gp130/CD130 Monoclonal specifically detects gp130/CD130 in Human samples. It is validated for Flow Cytometry.Specifications
gp130/CD130 | |
Monoclonal | |
PerCP | |
P40189 | |
IL6ST | |
Human recombinant gp130/CD130 | |
0.1 mL | |
Cancer, Immunology | |
3572 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry, Blocking Assay | |
MM0318-9X23 | |
Flow Cytometry | |
CD130, CD130 antigen, CDw130, DKFZp564F053, gp130, gp130 of the rheumatoid arthritis antigenic peptide-bearing soluble form, IL-6 receptor subunit beta, IL-6R subunit beta, IL-6RB, IL-6R-beta, interleukin 6 signal transducer (gp130, oncostatin M receptor), interleukin receptor beta chain, interleukin-6 receptor subunit beta, Interleukin-6 signal transducer, Membrane glycoprotein 130, membrane glycoprotein gp130, Oncostatin-M receptor subunit alpha | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction